Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000276.3 | NP_000267.2 | 20 - 21 | 2423 | 93 |
NM_001318784.1 | NP_001305713.1 | 20 - 21 | 2446 | 93 | |
NM_001587.3 | NP_001578.2 | 19 - 20 | 2399 | 93 | |
XM_005262422.2 | XP_005262479.1 | 19 - 20 | 2230 | 93 | |
XM_011531344.2 | XP_011529646.1 | 21 - 22 | 2335 | 93 | |
XM_011531345.2 | XP_011529647.1 | 21 - 22 | 2338 | 93 | |
XM_017029554.1 | XP_016885043.1 | 20 - 21 | 2354 | 93 | |
GenBank mRNA | AK226116.1 | - | 20 - 21 | 2446 | 93 |
AK293107.1 | - | 20 - 21 | 2422 | 93 | |
BC094726.1 | - | 19 - 20 | 2403 | 93 | |
BC130612.1 | - | 19 - 20 | 2343 | 93 | |
BC144106.1 | - | 19 - 20 | 2343 | 93 | |
U57627.2 | - | 20 - 21 | 2423 | 93 |