Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000276.3 | NP_000267.2 | 4 - 5 | 402 | 154 |
NM_001318784.1 | NP_001305713.1 | 4 - 5 | 425 | 154 | |
NM_001587.3 | NP_001578.2 | 4 - 5 | 402 | 154 | |
XM_005262422.2 | XP_005262479.1 | 3 - 4 | 209 | 154 | |
XM_017029554.1 | XP_016885043.1 | 4 - 5 | 333 | 154 | |
GenBank mRNA | AK226116.1 | - | 4 - 5 | 425 | 154 |
AK293107.1 | - | 4 - 5 | 401 | 154 | |
BC094726.1 | - | 4 - 5 | 406 | 154 | |
BC130612.1 | - | 4 - 5 | 346 | 154 | |
BC144106.1 | - | 4 - 5 | 346 | 154 | |
U57627.2 | - | 4 - 5 | 402 | 154 |