Assay Design



Gene Transcripts

Gene Symbol
C14orf37
Entrez Gene ID
145407
Gene Name
chromosome 14 open reading frame 37
Gene Aliases
c14_5376
Chromosome Location
Chr.14: 57993536 - 58152305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001872.3 NP_001001872.2 6 - 7 2339 63
XM_017021029.1 XP_016876518.1 6 - 7 2196 63
XM_017021030.1 XP_016876519.1 6 - 7 2339 63
GenBank mRNA AK292513.1 - 6 - 7 2313 63
AK292605.1 - 6 - 7 2256 63
AK294599.1 - 5 - 6 1550 63
BC062772.1 - 5 - 6 961 63
BC146861.1 - 6 - 7 2190 63
BX161411.1 - 6 - 7 2229 63
BX649130.1 - 5 - 6 1835 63

View Full Product Details