Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012469.3 | NP_036601.2 | 14 - 15 | 2080 | 70 |
XM_006723769.2 | XP_006723832.1 | 13 - 14 | 1870 | 70 | |
GenBank mRNA | AB019219.1 | - | 14 - 15 | 1994 | 70 |
AF026031.1 | - | 14 - 15 | 2011 | 70 | |
AF221842.1 | - | 14 - 15 | 2017 | 70 | |
AK314310.1 | - | 14 - 15 | 2004 | 70 | |
AL137320.1 | - | 5 - 6 | 711 | 70 | |
BC001666.2 | - | 14 - 15 | 1958 | 70 |