Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012469.3 | NP_036601.2 | 2 - 3 | 409 | 78 |
XM_006723769.2 | XP_006723832.1 | 2 - 3 | 418 | 78 | |
GenBank mRNA | AB019219.1 | - | 2 - 3 | 323 | 78 |
AF026031.1 | - | 2 - 3 | 340 | 78 | |
AF221842.1 | - | 2 - 3 | 346 | 78 | |
AK001554.1 | - | 2 - 3 | 352 | 78 | |
AK225993.1 | - | 2 - 3 | 363 | 78 | |
AK314310.1 | - | 2 - 3 | 333 | 78 | |
BC001666.2 | - | 2 - 3 | 287 | 78 | |
BP366197.1 | - | 2 - 3 | 409 | 78 |