Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012469.3 | NP_036601.2 | 8 - 9 | 1195 | 91 |
XM_006723769.2 | XP_006723832.1 | 8 - 9 | 1204 | 91 | |
GenBank mRNA | AB019219.1 | - | 8 - 9 | 1109 | 91 |
AF026031.1 | - | 8 - 9 | 1126 | 91 | |
AF221842.1 | - | 8 - 9 | 1132 | 91 | |
AK001554.1 | - | 8 - 9 | 1138 | 91 | |
AK225993.1 | - | 8 - 9 | 1149 | 91 | |
AK314310.1 | - | 8 - 9 | 1119 | 91 | |
BC001666.2 | - | 8 - 9 | 1073 | 91 |