Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018665.2 | NP_061135.2 | 14 - 15 | 2088 | 147 |
XM_011535926.1 | XP_011534228.1 | 13 - 14 | 1956 | 147 | |
XM_011535927.2 | XP_011534229.1 | 13 - 14 | 1545 | 147 | |
XM_011535928.2 | XP_011534230.1 | 13 - 14 | 1734 | 147 | |
XM_017011003.1 | XP_016866492.1 | 13 - 14 | 1674 | 147 | |
GenBank mRNA | AJ278110.1 | - | 14 - 15 | 1951 | 147 |
AK313686.1 | - | 14 - 15 | 1788 | 147 | |
AL136751.1 | - | 14 - 15 | 1776 | 147 | |
BC066938.1 | - | 14 - 15 | 1769 | 147 |