Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018665.2 | NP_061135.2 | 4 - 5 | 909 | 123 |
XM_011535927.2 | XP_011534229.1 | 3 - 4 | 366 | 123 | |
XM_011535928.2 | XP_011534230.1 | 3 - 4 | 555 | 123 | |
GenBank mRNA | AJ278110.1 | - | 4 - 5 | 772 | 123 |
AL136751.1 | - | 4 - 5 | 597 | 123 | |
BC066938.1 | - | 4 - 5 | 590 | 123 |