Assay Design



Gene Transcripts

Gene Symbol
MBD1
Entrez Gene ID
4152
Gene Name
methyl-CpG binding domain protein 1
Gene Aliases
CXXC3, PCM1, RFT
Chromosome Location
Chr.18: 50266882 - 50281774 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204136.1 NP_001191065.1 11 - 12 1581 76
NM_001204137.1 NP_001191066.1 12 - 13 1656 76
NM_001204139.1 NP_001191068.1 11 - 12 1581 76
NM_001204141.1 NP_001191070.1 10 - 11 1431 76
NM_001204142.1 NP_001191071.1 11 - 12 1581 76
NM_001204151.2 NP_001191080.1 10 - 11 1512 76
NM_001323942.1 NP_001310871.1 12 - 13 1656 76
NM_001323947.1 NP_001310876.1 12 - 13 1656 76
NM_001323949.1 NP_001310878.1 12 - 13 1648 76
NM_001323950.1 NP_001310879.1 11 - 12 1578 76
NM_001323951.1 NP_001310880.1 11 - 12 1581 76
NM_001323953.1 NP_001310882.1 10 - 11 1519 76
NM_015845.3 NP_056670.2 10 - 11 1512 76
NM_015846.3 NP_056671.2 11 - 12 1581 76
NM_015847.3 NP_056723.2 10 - 11 1434 76
XM_005258271.2 XP_005258328.1 11 - 12 1328 76
XM_006722456.2 XP_006722519.1 12 - 13 1403 76
XM_011525991.1 XP_011524293.1 12 - 13 1403 76
XM_011525993.2 XP_011524295.1 12 - 13 1403 76
XM_011525994.2 XP_011524296.1 12 - 13 1403 76
XM_011525998.2 XP_011524300.1 11 - 12 1325 76
XM_011525999.1 XP_011524301.1 11 - 12 1328 76
XM_011526002.1 XP_011524304.1 12 - 13 1403 76
XM_011526003.2 XP_011524305.1 12 - 13 1403 76
XM_011526007.1 XP_011524309.1 11 - 12 1328 76
XM_017025751.1 XP_016881240.1 12 - 13 1403 76
XM_017025752.1 XP_016881241.1 11 - 12 1328 76
XM_017025753.1 XP_016881242.1 11 - 12 1328 76
XM_017025754.1 XP_016881243.1 12 - 13 1403 76
XM_017025755.1 XP_016881244.1 11 - 12 1328 76
XM_017025756.1 XP_016881245.1 12 - 13 1403 76
XM_017025757.1 XP_016881246.1 12 - 13 1403 76
XM_017025758.1 XP_016881247.1 12 - 13 1403 76
XM_017025759.1 XP_016881248.1 12 - 13 1403 76
XM_017025760.1 XP_016881249.1 11 - 12 1328 76
XM_017025761.1 XP_016881250.1 12 - 13 1403 76
XM_017025762.1 XP_016881251.1 11 - 12 1328 76
XM_017025763.1 XP_016881252.1 11 - 12 1325 76
XM_017025764.1 XP_016881253.1 12 - 13 1403 76
XM_017025769.1 XP_016881258.1 11 - 12 1328 76
XM_017025777.1 XP_016881266.1 11 - 12 1328 76
GenBank mRNA AF078830.1 - 10 - 11 1144 76
AF078831.1 - 9 - 10 1075 76
AJ564845.1 - 11 - 12 1283 76
AK225889.1 - 11 - 12 1491 76
AK225948.1 - 11 - 12 1287 76
AK291519.1 - 11 - 12 1582 76
AK302004.1 - 12 - 13 1348 76
BC047353.1 - 10 - 11 1116 76
Y10746.1 - 10 - 11 1136 76

View Full Product Details