Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002437.4 | NP_002428.1 | 4 - 5 | 333 | 110 |
XM_005264326.3 | XP_005264383.1 | 4 - 5 | 345 | 110 | |
XM_006712021.3 | XP_006712084.1 | 5 - 6 | 485 | 110 | |
XM_017004150.1 | XP_016859639.1 | 5 - 6 | 3562 | 110 | |
XM_017004151.1 | XP_016859640.1 | 4 - 5 | 381 | 110 | |
XM_017004152.1 | XP_016859641.1 | 4 - 5 | 404 | 110 | |
GenBank mRNA | AF088036.1 | - | 4 - 5 | 304 | 110 |
AK093305.1 | - | 2 - 3 | 1339 | 110 | |
BC001115.2 | - | 3 - 4 | 324 | 110 | |
BC016289.2 | - | 3 - 4 | 267 | 110 | |
BI602840.1 | - | 4 - 5 | 334 | 110 | |
KU178195.1 | - | 3 - 4 | 277 | 110 | |
KU178196.1 | - | 3 - 4 | 242 | 110 | |
KU178197.1 | - | 3 - 4 | 277 | 110 | |
KU178198.1 | - | 2 - 3 | 161 | 110 | |
X76538.1 | - | 4 - 5 | 306 | 110 |