Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005066.2 | NP_005057.1 | 2 - 3 | 1104 | 141 |
NR_136702.1 | 2 - 3 | 1107 | 141 | ||
NR_136703.1 | 2 - 3 | 1107 | 141 | ||
XM_005271112.4 | XP_005271169.1 | 2 - 3 | 1123 | 141 | |
XM_005271113.4 | XP_005271170.1 | 2 - 3 | 1123 | 141 | |
XM_005271115.4 | XP_005271172.1 | 2 - 3 | 1123 | 141 | |
XM_011541950.2 | XP_011540252.1 | 2 - 3 | 1123 | 141 | |
XM_017002053.1 | XP_016857542.1 | 2 - 3 | 1123 | 141 | |
XM_017002054.1 | XP_016857543.1 | 2 - 3 | 1123 | 141 | |
GenBank mRNA | AK307985.1 | - | 3 - 4 | 912 | 142 |
BC004534.2 | - | 2 - 3 | 792 | 141 | |
BC027708.2 | - | 2 - 3 | 1095 | 141 | |
BC051192.1 | - | 2 - 3 | 1106 | 141 | |
X70944.1 | - | 2 - 3 | 1096 | 141 |