Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005066.2 | NP_005057.1 | 3 - 4 | 1411 | 69 |
NR_136702.1 | 3 - 4 | 1414 | 69 | ||
NR_136703.1 | 3 - 4 | 1414 | 69 | ||
XM_005271112.4 | XP_005271169.1 | 3 - 4 | 1430 | 69 | |
XM_005271113.4 | XP_005271170.1 | 3 - 4 | 1430 | 69 | |
XM_005271115.4 | XP_005271172.1 | 3 - 4 | 1430 | 69 | |
XM_011541950.2 | XP_011540252.1 | 3 - 4 | 1430 | 69 | |
XM_017002053.1 | XP_016857542.1 | 3 - 4 | 1430 | 69 | |
XM_017002054.1 | XP_016857543.1 | 3 - 4 | 1430 | 69 | |
GenBank mRNA | BC004534.2 | - | 3 - 4 | 1099 | 69 |
BC027708.2 | - | 3 - 4 | 1402 | 69 | |
BC051192.1 | - | 3 - 4 | 1413 | 69 | |
X16850.1 | - | 2 - 3 | 385 | 69 | |
X70944.1 | - | 3 - 4 | 1403 | 69 |