Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198844.2 | NP_942141.2 | 5 - 6 | 795 | 140 |
NM_199321.2 | NP_955353.1 | 6 - 7 | 861 | 140 | |
GenBank mRNA | AK292383.1 | - | 6 - 7 | 899 | 140 |
AY251603.1 | - | 6 - 7 | 844 | 140 | |
AY251604.1 | - | 5 - 6 | 778 | 140 | |
BC043152.2 | - | 5 - 6 | 781 | 140 |