Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000081.3 | NP_000072.2 | 18 - 19 | 5814 | 139 |
NM_001301365.1 | NP_001288294.1 | 18 - 19 | 6251 | 139 | |
XM_011544031.1 | XP_011542333.1 | 18 - 19 | 5814 | 139 | |
XM_011544032.1 | XP_011542334.1 | 18 - 19 | 6217 | 139 | |
XM_011544033.2 | XP_011542335.1 | 18 - 19 | 5964 | 139 | |
XM_011544034.1 | XP_011542336.1 | 18 - 19 | 5814 | 139 | |
XM_011544035.2 | XP_011542337.1 | 18 - 19 | 5814 | 139 | |
XM_011544036.2 | XP_011542338.1 | 14 - 15 | 3468 | 139 | |
XM_011544037.2 | XP_011542339.1 | 18 - 19 | 5814 | 139 | |
XM_011544039.2 | XP_011542341.1 | 18 - 19 | 5814 | 139 | |
XM_011544040.2 | XP_011542342.1 | 18 - 19 | 5814 | 139 | |
XM_017000150.1 | XP_016855639.1 | 18 - 19 | 5814 | 139 | |
XM_017000151.1 | XP_016855640.1 | 18 - 19 | 5814 | 139 | |
GenBank mRNA | BC144702.1 | - | 17 - 18 | 5688 | 139 |
U67615.1 | - | 18 - 19 | 5821 | 139 | |
U70064.1 | - | 18 - 19 | 6184 | 139 |