Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000081.3 | NP_000072.2 | 44 - 45 | 10327 | 102 |
NM_001301365.1 | NP_001288294.1 | 44 - 45 | 10764 | 102 | |
XM_011544031.1 | XP_011542333.1 | 45 - 46 | 10489 | 102 | |
XM_011544032.1 | XP_011542334.1 | 45 - 46 | 10892 | 102 | |
XM_011544033.2 | XP_011542335.1 | 45 - 46 | 10639 | 102 | |
XM_011544034.1 | XP_011542336.1 | 44 - 45 | 10351 | 102 | |
XM_011544036.2 | XP_011542338.1 | 41 - 42 | 8143 | 102 | |
GenBank mRNA | BX649113.1 | - | 13 - 14 | 5137 | 102 |
U67615.1 | - | 44 - 45 | 10334 | 102 |