Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167580.1 | NP_001161052.1 | 6 - 7 | 1078 | 66 |
NM_001323031.1 | NP_001309960.1 | 7 - 8 | 1611 | 66 | |
NM_001323032.1 | NP_001309961.1 | 7 - 8 | 1920 | 66 | |
NM_001323033.1 | NP_001309962.1 | 5 - 6 | 1587 | 66 | |
NM_001323034.1 | NP_001309963.1 | 7 - 8 | 1920 | 66 | |
NM_001323036.1 | NP_001309965.1 | 6 - 7 | 897 | 66 | |
NM_001323037.1 | NP_001309966.1 | 8 - 9 | 1827 | 66 | |
NM_001323038.1 | NP_001309967.1 | 7 - 8 | 1739 | 66 | |
NM_001323039.1 | NP_001309968.1 | 7 - 8 | 2012 | 66 | |
NM_001323040.1 | NP_001309969.1 | 7 - 8 | 1258 | 66 | |
NM_014848.5 | NP_055663.1 | 8 - 9 | 2008 | 66 | |
XM_005254998.3 | XP_005255055.1 | 7 - 8 | 1673 | 66 | |
XM_017022761.1 | XP_016878250.1 | 7 - 8 | 1624 | 66 | |
XM_017022762.1 | XP_016878251.1 | 7 - 8 | 1759 | 66 | |
GenBank mRNA | AB018278.1 | - | 6 - 7 | 1281 | 66 |
AF052188.1 | - | 7 - 8 | 1535 | 66 | |
AK294902.1 | - | 6 - 7 | 1080 | 66 | |
BC030011.2 | - | 8 - 9 | 1645 | 66 | |
CR457086.1 | - | 6 - 7 | 1117 | 66 |