Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195010.1 | NP_001181939.1 | 4 - 5 | 777 | 127 |
NM_021180.3 | NP_067003.2 | 4 - 5 | 673 | 127 | |
NM_198173.2 | NP_937816.1 | 4 - 5 | 842 | 127 | |
NM_198174.2 | NP_937817.3 | 4 - 5 | 842 | 127 | |
XM_011541869.1 | XP_011540171.1 | 4 - 5 | 723 | 127 | |
XM_011541870.2 | XP_011540172.1 | 3 - 4 | 644 | 127 | |
GenBank mRNA | AK074386.1 | - | 4 - 5 | 739 | 127 |
AK315164.1 | - | 4 - 5 | 644 | 127 | |
AY231160.1 | - | 4 - 5 | 673 | 127 | |
AY231161.1 | - | 3 - 4 | 486 | 127 | |
BC036890.2 | - | 4 - 5 | 788 | 127 |