Assay Design



Gene Transcripts

Gene Symbol
GRHL3
Entrez Gene ID
57822
Gene Name
grainyhead like transcription factor 3
Gene Aliases
SOM, TFCP2L4, VWS2
Chromosome Location
Chr.1: 24319322 - 24364482 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195010.1 NP_001181939.1 9 - 10 1371 74
NM_021180.3 NP_067003.2 9 - 10 1267 74
NM_198173.2 NP_937816.1 9 - 10 1436 74
NM_198174.2 NP_937817.3 9 - 10 1436 74
XM_011541869.1 XP_011540171.1 9 - 10 1317 74
XM_011541870.2 XP_011540172.1 8 - 9 1238 74
GenBank mRNA AK074386.1 - 9 - 10 1333 74
AY231160.1 - 9 - 10 1267 74
AY231161.1 - 8 - 9 1080 74
BC036890.2 - 9 - 10 1382 74

View Full Product Details