Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006653.4 | NP_006644.1 | 3 - 4 | 328 | 85 |
XM_011514254.2 | XP_011512556.1 | 3 - 4 | 1181 | 85 | |
XM_011514255.2 | XP_011512557.1 | 3 - 4 | 1176 | 85 | |
GenBank mRNA | AF036718.1 | - | 2 - 3 | 207 | 85 |
BC010611.1 | - | 2 - 3 | 146 | 85 | |
BM741175.1 | - | 3 - 4 | 315 | 85 | |
CR457026.1 | - | 1 - 2 | 71 | 85 | |
HY036161.1 | - | 3 - 4 | 85 |