Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006653.4 | NP_006644.1 | 5 - 6 | 677 | 79 |
XM_011514254.2 | XP_011512556.1 | 5 - 6 | 1530 | 79 | |
XM_011514255.2 | XP_011512557.1 | 5 - 6 | 1525 | 79 | |
XM_017010193.1 | XP_016865682.1 | 5 - 6 | 2285 | 79 | |
GenBank mRNA | AF036718.1 | - | 4 - 5 | 556 | 79 |
BC010611.1 | - | 4 - 5 | 495 | 79 | |
CR457026.1 | - | 3 - 4 | 420 | 79 |