Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034116.1 | NP_001029288.1 | 10 - 11 | 1159 | 89 |
NM_001318965.1 | NP_001305894.1 | 9 - 10 | 1118 | 89 | |
NM_001318966.1 | NP_001305895.1 | 10 - 11 | 1209 | 89 | |
NM_001318967.1 | NP_001305896.1 | 10 - 11 | 1152 | 89 | |
NM_001318968.1 | NP_001305897.1 | 9 - 10 | 1048 | 89 | |
NM_001318969.1 | NP_001305898.1 | 10 - 11 | 1133 | 89 | |
NM_015636.3 | NP_056451.3 | 10 - 11 | 1156 | 89 | |
NM_172195.3 | NP_751945.2 | 9 - 10 | 1115 | 89 | |
XM_006712132.1 | XP_006712195.1 | 10 - 11 | 1206 | 89 | |
XM_011533147.2 | XP_011531449.1 | 10 - 11 | 1014 | 89 | |
GenBank mRNA | AF057699.1 | - | 10 - 11 | 1026 | 89 |
AF218009.1 | - | 10 - 11 | 1181 | 89 | |
AJ011305.1 | - | 10 - 11 | 1032 | 89 | |
AJ011306.1 | - | 9 - 10 | 1115 | 89 | |
AK300734.1 | - | 9 - 10 | 1461 | 89 | |
AL050109.1 | - | 7 - 8 | 629 | 89 | |
BC001870.1 | - | 10 - 11 | 1017 | 89 | |
BC091502.1 | - | 10 - 11 | 1032 | 89 | |
JQ991007.1 | - | 10 - 11 | 1133 | 89 |