Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034116.1 | NP_001029288.1 | 12 - 13 | 1518 | 66 |
NM_001318965.1 | NP_001305894.1 | 11 - 12 | 1477 | 66 | |
NM_001318966.1 | NP_001305895.1 | 12 - 13 | 1568 | 66 | |
NM_001318967.1 | NP_001305896.1 | 12 - 13 | 1511 | 66 | |
NM_001318968.1 | NP_001305897.1 | 11 - 12 | 1407 | 66 | |
NM_001318969.1 | NP_001305898.1 | 12 - 13 | 1492 | 66 | |
NM_015636.3 | NP_056451.3 | 12 - 13 | 1515 | 66 | |
NM_172195.3 | NP_751945.2 | 11 - 12 | 1474 | 66 | |
XM_006712132.1 | XP_006712195.1 | 12 - 13 | 1565 | 66 | |
XM_011533147.2 | XP_011531449.1 | 12 - 13 | 1373 | 66 | |
GenBank mRNA | AF057699.1 | - | 12 - 13 | 1385 | 66 |
AF086520.1 | - | 3 - 4 | 402 | 66 | |
AF112207.1 | - | 12 - 13 | 1394 | 66 | |
AF218009.1 | - | 12 - 13 | 1539 | 66 | |
AJ011305.1 | - | 12 - 13 | 1391 | 66 | |
AJ011306.1 | - | 11 - 12 | 1474 | 66 | |
AL050109.1 | - | 9 - 10 | 988 | 66 | |
BC001870.1 | - | 12 - 13 | 1376 | 66 | |
BC091502.1 | - | 12 - 13 | 1391 | 66 | |
JQ991007.1 | - | 12 - 13 | 1492 | 66 |