Assay Design
Gene Transcripts
- Gene Symbol
- NPEPL1
- Entrez Gene ID
-
79716
- Gene Name
- aminopeptidase-like 1
- Gene Aliases
-
bA261P9.2
- Chromosome Location
-
Chr.20: 58689131 - 58715844 on Build GRCh38
- Gene Symbol
- STX16-NPEPL1
- Entrez Gene ID
-
100534593
- Gene Name
- STX16-NPEPL1 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 58651253 - 58715844 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037945.1 |
|
19 - 20 |
2960 |
57 |
GenBank mRNA |
AB209296.1 |
- |
19 - 20 |
2779 |
57 |
View Full Product Details