Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001093771.2 | NP_001087240.1 | 8 - 9 | 895 | 143 |
NM_001261445.1 | NP_001248374.1 | 6 - 7 | 1317 | 143 | |
NM_001261446.1 | NP_001248375.1 | 5 - 6 | 872 | 143 | |
NM_003330.3 | NP_003321.3 | 6 - 7 | 1232 | 143 | |
NM_182729.2 | NP_877393.1 | 6 - 7 | 995 | 143 | |
NM_182742.2 | NP_877419.1 | 5 - 6 | 1122 | 143 | |
NM_182743.2 | NP_877420.1 | 5 - 6 | 885 | 143 | |
GenBank mRNA | AF077367.1 | - | 5 - 6 | 549 | 143 |
AF208018.1 | - | 4 - 5 | 445 | 143 | |
AJ001050.1 | - | 4 - 5 | 458 | 143 | |
AK128515.1 | - | 5 - 6 | 780 | 143 | |
AK226071.1 | - | 6 - 7 | 703 | 143 | |
AK293322.1 | - | 5 - 6 | 532 | 143 | |
AK295067.1 | - | 6 - 7 | 631 | 143 | |
AK296495.1 | - | 5 - 6 | 522 | 143 | |
AK304241.1 | - | 8 - 9 | 878 | 143 | |
AK308881.1 | - | 6 - 7 | 926 | 143 | |
AK312262.1 | - | 6 - 7 | 894 | 143 | |
AY057105.1 | - | 8 - 9 | 871 | 143 | |
BC018122.1 | - | 6 - 7 | 608 | 143 | |
BT019640.1 | - | 4 - 5 | 421 | 143 | |
CR536506.1 | - | 4 - 5 | 421 | 143 | |
D88687.1 | - | 6 - 7 | 673 | 143 | |
S79851.1 | - | 6 - 7 | 860 | 143 | |
X91247.1 | - | 6 - 7 | 860 | 143 |