Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324511.1 | NP_001311440.1 | 1 - 2 | 461 | 90 |
NM_001324512.1 | NP_001311441.1 | 1 - 2 | 461 | 90 | |
NM_030955.3 | NP_112217.2 | 1 - 2 | 461 | 90 | |
XM_017009905.1 | XP_016865394.1 | 1 - 2 | 449 | 90 | |
GenBank mRNA | AJ250725.1 | - | 1 - 2 | 451 | 90 |
AY358745.1 | - | 1 - 2 | 244 | 90 | |
BC058841.1 | - | 1 - 2 | 292 | 90 | |
BC131733.1 | - | 1 - 2 | 210 | 90 | |
BC139900.1 | - | 1 - 2 | 132 | 90 | |
BG108002.1 | - | 1 - 2 | 338 | 90 |