Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324511.1 | NP_001311440.1 | 2 - 3 | 823 | 87 |
NM_001324512.1 | NP_001311441.1 | 2 - 3 | 823 | 87 | |
NM_030955.3 | NP_112217.2 | 2 - 3 | 823 | 87 | |
XM_017009905.1 | XP_016865394.1 | 2 - 3 | 811 | 87 | |
GenBank mRNA | AJ250725.1 | - | 2 - 3 | 813 | 87 |
AY358745.1 | - | 2 - 3 | 606 | 87 | |
BC058841.1 | - | 2 - 3 | 654 | 87 | |
BC131733.1 | - | 2 - 3 | 572 | 87 | |
BC139900.1 | - | 2 - 3 | 494 | 87 | |
BQ774480.1 | - | 1 - 3 | 87 | ||
N93301.1 | - | 1 - 2 | 87 |