Assay Design



Gene Transcripts

Gene Symbol
PDLIM2
Entrez Gene ID
64236
Gene Name
PDZ and LIM domain 2
Gene Aliases
MYSTIQUE, SLIM
Chromosome Location
Chr.8: 22578741 - 22598025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021630.5 NP_067643.3 1 - 2 782 52
GenBank mRNA AF289563.1 - 1 - 2 382 52
AK027800.1 - 1 - 2 393 52
AK092968.1 - 1 - 2 782 52
BC021556.1 - 1 - 2 263 52
BC071774.1 - 1 - 2 393 52

View Full Product Details