Assay Design



Gene Transcripts

Gene Symbol
PDLIM2
Entrez Gene ID
64236
Gene Name
PDZ and LIM domain 2
Gene Aliases
MYSTIQUE, SLIM
Chromosome Location
Chr.8: 22578741 - 22598025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021630.5 NP_067643.3 7 - 8 1412 91
NM_176871.3 NP_789847.1 6 - 7 754 91
NM_198042.3 NP_932159.1 6 - 7 754 91
GenBank mRNA AK024498.1 - 6 - 7 697 91
AK027800.1 - 7 - 8 1023 91
AK056748.1 - 1 - 2 1179 91
AK092968.1 - 7 - 8 1412 91
AY007729.1 - 5 - 6 697 91
AY070438.1 - 6 - 7 778 91
AY217349.1 - 6 - 7 739 91
BC021556.1 - 7 - 8 893 91
BC071774.1 - 7 - 8 1023 91
DN997519.1 - 7 - 8 698 91

View Full Product Details