Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242935.2 | NP_001229864.1 | 9 - 10 | 2032 | 113 |
NM_001308100.1 | NP_001295029.1 | 13 - 14 | 4529 | 113 | |
XM_005272138.3 | XP_005272195.1 | 11 - 12 | 3007 | 113 | |
XM_005272139.1 | XP_005272196.1 | 11 - 12 | 2947 | 113 | |
XM_006714734.2 | XP_006714797.1 | 11 - 12 | 3140 | 113 | |
XM_011543737.2 | XP_011542039.1 | 12 - 13 | 3142 | 113 | |
XM_011543738.2 | XP_011542040.1 | 12 - 13 | 3141 | 113 | |
XM_011543739.1 | XP_011542041.1 | 12 - 13 | 3067 | 113 | |
XM_011543741.2 | XP_011542043.1 | 12 - 13 | 3091 | 113 | |
XM_011543743.2 | XP_011542045.1 | 12 - 13 | 3516 | 113 | |
XM_011543745.1 | XP_011542047.1 | 11 - 12 | 2971 | 113 | |
XM_011543749.2 | XP_011542051.1 | 9 - 10 | 2322 | 113 | |
XM_011543750.1 | XP_011542052.1 | 8 - 9 | 1800 | 113 | |
XM_017010078.1 | XP_016865567.1 | 12 - 13 | 5469 | 113 | |
XM_017010083.1 | XP_016865572.1 | 9 - 10 | 1948 | 113 | |
GenBank mRNA | AK001617.1 | - | 7 - 8 | 1768 | 113 |
BC040552.1 | - | 13 - 14 | 4391 | 113 | |
BC094759.1 | - | 9 - 10 | 1910 | 113 | |
DQ227317.1 | - | 9 - 10 | 1976 | 113 |