Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256317.1 | NP_001243246.1 | 2 - 3 | 298 | 101 |
NM_024022.2 | NP_076927.1 | 2 - 3 | 298 | 101 | |
NM_032405.1 | NP_115781.1 | 2 - 3 | 298 | 101 | |
NR_046020.1 | 2 - 3 | 1053 | 101 | ||
GenBank mRNA | AB038157.1 | - | 2 - 3 | 298 | 101 |
AB038160.1 | - | 2 - 3 | 298 | 101 | |
AF201380.1 | - | 2 - 3 | 240 | 101 | |
AF267741.1 | - | 2 - 3 | 262 | 101 | |
AK172842.1 | - | 2 - 3 | 261 | 101 | |
AY358458.1 | - | 2 - 3 | 222 | 101 | |
AY633572.1 | - | 2 - 3 | 1053 | 101 | |
BC074846.2 | - | 1 - 2 | 126 | 101 | |
BC074847.2 | - | 1 - 2 | 126 | 101 |