Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276725.1 | NP_001263654.1 | 2 - 3 | 434 | 172 |
NM_001276726.1 | NP_001263655.1 | 2 - 3 | 314 | 172 | |
NM_001276727.1 | NP_001263656.1 | 2 - 3 | 265 | 172 | |
GenBank mRNA | AK095997.1 | - | 2 - 3 | 290 | 172 |
BC009561.1 | - | 2 - 3 | 239 | 172 | |
BQ018679.1 | - | 2 - 3 | 405 | 172 | |
CD674194.1 | - | 2 - 3 | 197 | 172 | |
DQ121386.1 | - | 2 - 3 | 247 | 172 |