Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006657.1 | NP_001006658.1 | 23 - 24 | 2806 | 110 |
NM_020779.3 | NP_065830.2 | 22 - 23 | 2773 | 110 | |
XM_011533007.2 | XP_011531309.1 | 12 - 13 | 1498 | 110 | |
GenBank mRNA | AB037757.2 | - | 23 - 24 | 2814 | 110 |
AK122917.1 | - | 22 - 23 | 2698 | 110 | |
BC036659.2 | - | 22 - 23 | 2772 | 110 |