Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006657.1 | NP_001006658.1 | 6 - 7 | 678 | 128 |
NM_020779.3 | NP_065830.2 | 6 - 7 | 678 | 128 | |
GenBank mRNA | AB037757.2 | - | 6 - 7 | 686 | 128 |
AK091205.1 | - | 1 - 2 | 107 | 128 | |
AK122917.1 | - | 6 - 7 | 603 | 128 | |
AK315094.1 | - | 6 - 7 | 603 | 128 | |
BC036659.2 | - | 6 - 7 | 677 | 128 |