Assay Design



Gene Transcripts

Gene Symbol
TM2D2
Entrez Gene ID
83877
Gene Name
TM2 domain containing 2
Gene Aliases
BLP1
Chromosome Location
Chr.8: 38988808 - 38997139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024380.1 NP_001019551.1 1 - 2 96 105

View Full Product Details