Assay Design



Gene Transcripts

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
-
Chromosome Location
Chr.9: 111686175 - 111795949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_109816.1 1 - 2 110 162
XM_006716974.2 XP_006717037.1 1 - 2 94 162
XM_006716975.2 XP_006717038.1 1 - 2 94 162
XM_011518309.1 XP_011516611.1 1 - 2 94 162
XM_017014340.1 XP_016869829.1 1 - 2 1051 162
XM_017014341.1 XP_016869830.1 1 - 2 94 162
GenBank mRNA AK057341.1 - 1 - 2 45 162
AL704322.1 - 1 - 2 111 162
BC112357.1 - 1 - 2 38 162
BC144680.1 - 1 - 2 34 162

View Full Product Details