Assay Design



Gene Transcripts

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
-
Chromosome Location
Chr.9: 111686175 - 111795949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173521.4 NP_775792.4 2 - 3 194 127
NR_109816.1 4 - 5 396 127
XM_006716974.2 XP_006717037.1 4 - 5 380 127
XM_006716975.2 XP_006717038.1 4 - 5 380 127
XM_011518302.2 XP_011516604.1 2 - 3 2189 127
XM_011518303.1 XP_011516605.1 2 - 3 161 127
XM_011518309.1 XP_011516611.1 4 - 5 380 127
XM_017014341.1 XP_016869830.1 4 - 5 380 127
GenBank mRNA AK057341.1 - 4 - 5 331 127
AL704322.1 - 4 - 5 397 127
AL833535.1 - 2 - 3 189 127
BC112357.1 - 4 - 5 324 127
BC144680.1 - 4 - 5 320 127
DB066382.1 - 2 - 3 194 127

View Full Product Details