Assay Design



Gene Transcripts

Gene Symbol
COPS2
Entrez Gene ID
9318
Gene Name
COP9 signalosome subunit 2
Gene Aliases
ALIEN, CSN2, SGN2, TRIP15
Chromosome Location
Chr.15: 49125274 - 49155657 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143887.1 NP_001137359.1 9 - 10 1041 127
NM_004236.3 NP_004227.1 9 - 10 1020 127
GenBank mRNA AB209799.1 - 9 - 10 967 127
AF084260.1 - 9 - 10 956 127
AF100762.1 - 9 - 10 946 127
AF120268.1 - 9 - 10 931 126
AF212227.1 - 9 - 10 967 127
AK222590.1 - 9 - 10 944 127
AK295598.1 - 7 - 8 775 127
AK312643.1 - 9 - 10 956 127
BC012629.1 - 9 - 10 957 127
CR542063.1 - 9 - 10 941 127

View Full Product Details