Assay Design



Gene Transcripts

Gene Symbol
TCF12
Entrez Gene ID
6938
Gene Name
transcription factor 12
Gene Aliases
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
Chromosome Location
Chr.15: 56918090 - 57291261 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322151.1 NP_001309080.1 8 - 9 863 133
NM_001322152.1 NP_001309081.1 8 - 9 868 133
NM_001322154.1 NP_001309083.1 8 - 9 863 133
NM_001322156.1 NP_001309085.1 9 - 10 930 133
NM_001322157.1 NP_001309086.1 8 - 9 868 133
NM_001322158.1 NP_001309087.1 9 - 10 930 133
NM_001322159.1 NP_001309088.1 8 - 9 868 133
NM_001322161.1 NP_001309090.1 8 - 9 714 133
NM_001322162.1 NP_001309091.1 8 - 9 714 133
NM_001322164.1 NP_001309093.1 8 - 9 733 133
NM_001322165.1 NP_001309094.1 7 - 8 697 133
NM_003205.3 NP_003196.1 8 - 9 868 133
NM_207036.1 NP_996919.1 8 - 9 858 133
NM_207037.1 NP_996920.1 8 - 9 868 133
NM_207038.1 NP_996921.1 8 - 9 858 133
XM_011521959.2 XP_011520261.1 9 - 10 864 133
XM_011521960.2 XP_011520262.1 9 - 10 864 133
XM_011521961.2 XP_011520263.1 9 - 10 864 133
XM_011521962.2 XP_011520264.1 9 - 10 864 133
XM_011521963.2 XP_011520265.1 9 - 10 864 133
XM_011521965.1 XP_011520267.1 8 - 9 715 133
XM_011521967.1 XP_011520269.1 8 - 9 715 133
XM_017022520.1 XP_016878009.1 8 - 9 834 133
GenBank mRNA AK294617.1 - 8 - 9 816 133
AK302923.1 - 8 - 9 818 133
AK312710.1 - 8 - 9 828 133
AL831981.2 - 8 - 9 836 133
BC050556.1 - 8 - 9 809 133
M80627.1 - 8 - 9 787 133
M83233.1 - 8 - 9 662 133

View Full Product Details