Assay Design



Gene Transcripts

Gene Symbol
BTBD1
Entrez Gene ID
53339
Gene Name
BTB domain containing 1
Gene Aliases
C15orf1, NS5ATP8
Chromosome Location
Chr.15: 83016423 - 83067434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011885.1 NP_001011885.1 3 - 4 867 141
NM_025238.3 NP_079514.1 3 - 4 867 141
GenBank mRNA AF257241.1 - 3 - 4 746 141
AF355402.1 - 3 - 4 747 141
AF529369.1 - 3 - 4 664 141
AK000731.1 - 3 - 4 731 141
AK225355.1 - 3 - 4 731 141
AK225362.1 - 3 - 4 731 141
AK294410.1 - 1 - 2 119 141
AL136853.1 - 3 - 4 747 141
AY364253.1 - 3 - 4 664 141
BC017464.1 - 3 - 4 694 141
BC028097.1 - 3 - 4 767 141

View Full Product Details