Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018590.4 | NP_061060.3 | 1 - 2 | 123 | 142 |
NR_135077.1 | 1 - 2 | 123 | 142 | ||
XM_011539938.1 | XP_011538240.1 | 1 - 2 | 123 | 142 | |
XM_017016388.1 | XP_016871877.1 | 1 - 2 | 123 | 142 | |
XM_017016389.1 | XP_016871878.1 | 1 - 2 | 123 | 142 | |
XM_017016390.1 | XP_016871879.1 | 1 - 2 | 123 | 142 | |
XM_017016391.1 | XP_016871880.1 | 1 - 2 | 123 | 142 | |
GenBank mRNA | AB090811.1 | - | 1 - 2 | 82 | 142 |
AK125300.1 | - | 1 - 2 | 35 | 142 | |
BC030268.1 | - | 1 - 2 | 82 | 142 | |
BX640967.1 | - | 1 - 2 | 62 | 142 | |
DA674683.1 | - | 1 - 2 | 118 | 142 | |
DB291068.1 | - | 1 - 2 | 123 | 142 |