Assay Design



Gene Transcripts

Gene Symbol
PRKAG3
Entrez Gene ID
53632
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 3
Gene Aliases
AMPKG3
Chromosome Location
Chr.2: 218821853 - 218831803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017431.2 NP_059127.2 1 - 2 51 103
XM_017004343.1 XP_016859832.1 1 - 2 65 103
GenBank mRNA AJ249977.1 - 1 - 2 53 103
AK300078.1 - 1 - 2 63 103
AK300687.1 - 1 - 2 65 103
AK309865.1 - 1 - 2 65 103
BC098102.1 - 1 - 2 51 103
BC098255.1 - 1 - 2 51 103
BC098277.1 - 1 - 2 51 103
BC098306.1 - 1 - 2 51 103

View Full Product Details