Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282983.1 | NP_001269912.1 | 11 - 12 | 1266 | 146 |
NM_001282984.1 | NP_001269913.1 | 11 - 12 | 1266 | 146 | |
NM_014808.3 | NP_055623.1 | 11 - 12 | 1266 | 146 | |
XM_005247050.3 | XP_005247107.1 | 11 - 12 | 1230 | 146 | |
XM_011512233.1 | XP_011510535.1 | 11 - 12 | 1230 | 146 | |
XM_011512234.1 | XP_011510536.1 | 11 - 12 | 1155 | 146 | |
XM_011512235.1 | XP_011510537.1 | 12 - 13 | 1488 | 146 | |
XM_011512236.1 | XP_011510538.1 | 11 - 12 | 1284 | 146 | |
XM_017005416.1 | XP_016860905.1 | 12 - 13 | 1445 | 146 | |
XM_017005417.1 | XP_016860906.1 | 12 - 13 | 1445 | 146 | |
XM_017005418.1 | XP_016860907.1 | 9 - 10 | 1083 | 146 | |
XM_017005419.1 | XP_016860908.1 | 11 - 12 | 1230 | 146 | |
XM_017005420.1 | XP_016860909.1 | 11 - 12 | 1230 | 146 | |
GenBank mRNA | AB018336.1 | - | 11 - 12 | 1219 | 146 |
BC021301.2 | - | 11 - 12 | 1218 | 146 |