Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017962.2 | NP_001017962.1 | 8 - 9 | 1314 | 125 |
NM_001142595.1 | NP_001136067.1 | 9 - 10 | 1407 | 125 | |
GenBank mRNA | BC034998.1 | - | 8 - 9 | 1207 | 125 |
CD013930.1 | - | 9 - 10 | 1286 | 125 | |
M24487.1 | - | 8 - 9 | 1191 | 125 |