Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079520.1 | NP_001072988.1 | 2 - 3 | 643 | 65 |
NM_016651.5 | NP_057735.2 | 2 - 3 | 643 | 65 | |
NR_046093.1 | 2 - 3 | 252 | 65 | ||
NR_046095.1 | 2 - 3 | 560 | 65 | ||
XM_006720167.3 | XP_006720230.1 | 2 - 3 | 580 | 65 | |
GenBank mRNA | AK299879.1 | - | 2 - 3 | 560 | 65 |
AK316164.1 | - | 2 - 3 | 350 | 65 | |
AK316237.1 | - | 2 - 3 | 252 | 65 | |
AK316423.1 | - | 1 - 2 | 119 | 65 |