Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142276.1 | NP_001135748.1 | 3 - 4 | 559 | 69 |
NM_001142277.1 | NP_001135749.1 | 3 - 4 | 559 | 69 | |
NM_001243299.1 | NP_001230228.1 | 3 - 4 | 477 | 69 | |
NM_001642.2 | NP_001633.1 | 3 - 4 | 559 | 69 | |
GenBank mRNA | AK225283.1 | - | 3 - 4 | 486 | 69 |
AK304736.1 | - | 3 - 4 | 468 | 69 | |
BC000373.2 | - | 3 - 4 | 459 | 69 | |
L09209.1 | - | 3 - 4 | 474 | 69 | |
L19597.1 | - | 3 - 4 | 470 | 69 | |
L23114.1 | - | 2 - 3 | 290 | 69 | |
L27631.1 | - | 3 - 4 | 475 | 69 | |
S60099.1 | - | 3 - 4 | 474 | 69 | |
Z22572.1 | - | 3 - 4 | 469 | 69 |