Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014808.3 | NP_055623.1 | 17 - 18 | 2050 | 89 |
XM_005247050.3 | XP_005247107.1 | 17 - 18 | 2014 | 89 | |
XM_011512233.1 | XP_011510535.1 | 17 - 18 | 2014 | 89 | |
XM_011512234.1 | XP_011510536.1 | 17 - 18 | 1939 | 89 | |
XM_011512235.1 | XP_011510537.1 | 18 - 19 | 2272 | 89 | |
XM_011512236.1 | XP_011510538.1 | 17 - 18 | 2068 | 89 | |
XM_017005416.1 | XP_016860905.1 | 18 - 19 | 2229 | 89 | |
XM_017005417.1 | XP_016860906.1 | 18 - 19 | 2229 | 89 | |
XM_017005418.1 | XP_016860907.1 | 15 - 16 | 1867 | 89 | |
GenBank mRNA | AB018336.1 | - | 17 - 18 | 2003 | 89 |