Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014808.3 | NP_055623.1 | 26 - 27 | 3217 | 97 |
XM_011512233.1 | XP_011510535.1 | 27 - 28 | 3202 | 97 | |
XM_011512234.1 | XP_011510536.1 | 27 - 28 | 3127 | 97 | |
XM_011512235.1 | XP_011510537.1 | 28 - 29 | 3460 | 97 | |
XM_011512236.1 | XP_011510538.1 | 27 - 28 | 3256 | 97 | |
XM_017005416.1 | XP_016860905.1 | 28 - 29 | 3417 | 97 | |
XM_017005417.1 | XP_016860906.1 | 27 - 28 | 3396 | 97 | |
XM_017005418.1 | XP_016860907.1 | 24 - 25 | 3034 | 97 | |
GenBank mRNA | AB018336.1 | - | 26 - 27 | 3170 | 97 |
AK127510.1 | - | 5 - 6 | 2448 | 97 | |
AL122052.1 | - | 5 - 6 | 582 | 97 |