Assay Design



Gene Transcripts

Gene Symbol
DOK5
Entrez Gene ID
55816
Gene Name
docking protein 5
Gene Aliases
C20orf180, IRS-6, IRS6
Chromosome Location
Chr.20: 54475472 - 54651171 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018431.4 NP_060901.2 4 - 5 888 149
NM_177959.2 NP_808874.1 4 - 5 534 149
XM_011528904.1 XP_011527206.1 4 - 5 517 149
GenBank mRNA AF132732.1 - 4 - 5 610 149
AF466368.1 - 4 - 5 413 149
AK313222.1 - 4 - 5 600 149
AL050069.1 - 3 - 4 291 149
BC008992.1 - 4 - 5 534 149
BI758267.1 - 4 - 5 633 150

View Full Product Details