Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003930.4 | NP_003921.2 | 1 - 2 | 376 | 153 |
XM_017012771.1 | XP_016868260.1 | 1 - 2 | 338 | 153 | |
GenBank mRNA | AB014486.1 | - | 1 - 2 | 95 | 153 |
AF051323.1 | - | 1 - 2 | 234 | 153 | |
AK222885.1 | - | 1 - 2 | 222 | 153 | |
AK313209.1 | - | 1 - 2 | 222 | 153 | |
BC002893.2 | - | 1 - 2 | 220 | 153 | |
BC036044.1 | - | 1 - 2 | 223 | 153 | |
HY044523.1 | - | 1 - 2 | 377 | 153 |