Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258415.1 | NP_001245344.1 | 10 - 11 | 1428 | 123 |
NM_001258416.1 | NP_001245345.1 | 11 - 12 | 1314 | 123 | |
NM_001258417.1 | NP_001245346.1 | 10 - 11 | 1128 | 123 | |
NM_001258418.1 | NP_001245347.1 | 10 - 11 | 1246 | 123 | |
NM_020809.3 | NP_065860.2 | 11 - 12 | 1425 | 123 | |
XM_006718893.3 | XP_006718956.1 | 10 - 11 | 1334 | 123 | |
GenBank mRNA | AB037812.1 | - | 10 - 11 | 1150 | 123 |
AK307602.1 | - | 10 - 11 | 1334 | 123 | |
AY496263.1 | - | 11 - 12 | 1425 | 123 | |
AY496264.1 | - | 10 - 11 | 1128 | 123 | |
AY496265.1 | - | 10 - 11 | 1246 | 123 | |
AY496266.1 | - | 10 - 11 | 1428 | 123 | |
AY496267.1 | - | 11 - 12 | 1314 | 123 |